Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCCAAGGAGATGGGGCTCTTCCT[G/T]CCCTTTCAGATAGTTCCAGGGGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
25 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMCC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMCC2 - transmembrane and coiled-coil domain family 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242925.1 | Intron | NP_001229854.1 | ||||
NM_001297611.1 | Intron | NP_001284540.1 | ||||
NM_001297613.1 | Intron | NP_001284542.1 | ||||
NM_014858.3 | Intron | NP_055673.2 | ||||
XM_005245684.1 | Intron | XP_005245741.1 | ||||
XM_005245685.4 | Intron | XP_005245742.1 | ||||
XM_005245686.3 | Intron | XP_005245743.1 | ||||
XM_017003000.1 | Intron | XP_016858489.1 |