Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGTTGCATCATTGTGAGGAAAAC[A/C]ACTTAGTATTTTATAGTGAGGTGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PNLIPRP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PNLIPRP3 - pancreatic lipase related protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011709.2 | 54 | UTR 5 | NP_001011709.2 | |||
XM_011539276.1 | 54 | UTR 5 | XP_011537578.1 | |||
XM_011539277.1 | 54 | Intron | XP_011537579.1 | |||
XM_011539278.1 | 54 | Intron | XP_011537580.1 | |||
XM_011539279.1 | 54 | UTR 5 | XP_011537581.1 |