Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTTGGTTAGTTCATTTTAGGGAG[A/C]TCTGAAGGCAAAGTCACAGTAGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616670 MIM: 602145 MIM: 613315 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESYT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ESYT1 - extended synaptotagmin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PA2G4 - proliferation-associated 2G4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL41 - ribosomal protein L41 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZC3H10 - zinc finger CCCH-type containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303124.1 | Intron | NP_001290053.1 | ||||
NM_001303125.1 | Intron | NP_001290054.1 | ||||
NM_032786.2 | Intron | NP_116175.1 |