Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608255 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRAF3IP3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TRAF3IP3 - TRAF3 interacting protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287754.1 | Intron | NP_001274683.1 | ||||
NM_001320143.1 | Intron | NP_001307072.1 | ||||
NM_001320144.1 | Intron | NP_001307073.1 | ||||
NM_025228.3 | Intron | NP_079504.2 | ||||
XM_005273279.4 | Intron | XP_005273336.1 | ||||
XM_005273280.2 | Intron | XP_005273337.1 | ||||
XM_011510018.2 | Intron | XP_011508320.1 | ||||
XM_011510019.1 | Intron | XP_011508321.1 | ||||
XM_017002399.1 | Intron | XP_016857888.1 | ||||
XM_017002400.1 | Intron | XP_016857889.1 |
Set Membership: |
HapMap |