Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGGTAAAATCCTGCACTTAGGTT[C/T]CCTCTCTGGTTGATCTTTGAGGGCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 157680 MIM: 609372 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC25C PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CDC25C - cell division cycle 25C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM53C - family with sequence similarity 53 member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135647.1 | Intron | NP_001129119.1 | ||||
NM_016605.2 | Intron | NP_057689.1 |