Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCTGTCCACCCTTGCCCCAGGAG[A/G]GCCACAGGGAAGCATCCATCAGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607875 MIM: 600174 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INPP5K PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INPP5K - inositol polyphosphate-5-phosphatase K | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PITPNA - phosphatidylinositol transfer protein alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006224.3 | 2583 | UTR 3 | NP_006215.1 | |||
XM_011523939.1 | 2583 | UTR 3 | XP_011522241.1 | |||
XM_011523940.1 | 2583 | UTR 3 | XP_011522242.1 |
PITPNA-AS1 - PITPNA antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |