Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAGAAAAAGGAATAGATCTTTCT[A/G]AAACCTGGCCTGCACTTAGAGCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603054 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GREM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GREM1 - gremlin 1, DAN family BMP antagonist | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001191322.1 | Intron | NP_001178251.1 | ||||
NM_001191323.1 | Intron | NP_001178252.1 | ||||
NM_013372.6 | Intron | NP_037504.1 | ||||
XM_005254301.2 | Intron | XP_005254358.1 | ||||
XM_017022077.1 | Intron | XP_016877566.1 |
LOC100131315 - uncharacterized LOC100131315 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |