Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACCGCTCAGGCCGCGAGCAGCAG[A/C]AAGGCGCACCGCCACTGTCGCCGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606667 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LGR5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LGR5 - leucine rich repeat containing G protein-coupled receptor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277226.1 | 59 | UTR 5 | NP_001264155.1 | |||
NM_001277227.1 | 59 | UTR 5 | NP_001264156.1 | |||
NM_003667.3 | 59 | UTR 5 | NP_003658.1 |