Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611485 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP4F12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CYP4F12 - cytochrome P450 family 4 subfamily F member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023944.3 | Intron | NP_076433.3 | ||||
XM_006722850.3 | Intron | XP_006722913.1 | ||||
XM_011528202.2 | Intron | XP_011526504.1 | ||||
XM_011528203.2 | Intron | XP_011526505.1 | ||||
XM_011528204.2 | Intron | XP_011526506.1 | ||||
XM_011528205.2 | Intron | XP_011526507.1 | ||||
XM_011528207.2 | Intron | XP_011526509.1 | ||||
XM_011528208.2 | Intron | XP_011526510.1 | ||||
XM_017027172.1 | Intron | XP_016882661.1 |
Set Membership: |
HapMap |