Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAGAAAACATGGGTACATTGCCA[C/T]TCTACCAATGATGGTATTACCTATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616337 MIM: 608126 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SOCS4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SOCS4 - suppressor of cytokine signaling 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080867.2 | Intron | NP_543143.1 | ||||
NM_199421.1 | Intron | NP_955453.1 | ||||
XM_011536425.1 | Intron | XP_011534727.1 | ||||
XM_011536426.1 | Intron | XP_011534728.1 |
WDHD1 - WD repeat and HMG-box DNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |