Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTAGAGGATTGCGGTTTCCCTTCA[G/T]CTCCGCGGCTCTTATTCCTCCCCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600438 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TFAM PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TFAM - transcription factor A, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270782.1 | 150 | UTR 5 | NP_001257711.1 | |||
NM_003201.2 | 150 | UTR 5 | NP_003192.1 | |||
XM_011540120.2 | 150 | Intron | XP_011538422.1 | |||
XM_011540121.2 | 150 | Intron | XP_011538423.1 |