Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAAACTGGGGAAGACAGAAGTCA[C/T]AGTCAGAAGGAACTAGCTGTTTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609829 MIM: 606105 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FSD1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FSD1L - fibronectin type III and SPRY domain containing 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145313.2 | Intron | NP_001138785.1 | ||||
NM_001287191.1 | Intron | NP_001274120.1 | ||||
NM_001287192.1 | Intron | NP_001274121.1 | ||||
NM_031919.4 | Intron | NP_114125.1 | ||||
XM_005252254.2 | Intron | XP_005252311.1 | ||||
XM_005252257.3 | Intron | XP_005252314.1 | ||||
XM_005252260.3 | Intron | XP_005252317.1 | ||||
XM_011519077.2 | Intron | XP_011517379.1 | ||||
XM_011519078.2 | Intron | XP_011517380.2 | ||||
XM_011519079.2 | Intron | XP_011517381.2 | ||||
XM_011519080.2 | Intron | XP_011517382.1 | ||||
XM_011519081.2 | Intron | XP_011517383.1 | ||||
XM_017015182.1 | Intron | XP_016870671.1 | ||||
XM_017015183.1 | Intron | XP_016870672.1 | ||||
XM_017015184.1 | Intron | XP_016870673.1 | ||||
XM_017015185.1 | Intron | XP_016870674.1 | ||||
XM_017015186.1 | Intron | XP_016870675.1 | ||||
XM_017015187.1 | Intron | XP_016870676.1 |
SLC44A1 - solute carrier family 44 member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |