Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCTCTTTTTTATTTTTGCTTTTC[C/T]AGTGGATTTGGCAGCTAATTCACTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607663 MIM: 610693 MIM: 616283 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX25 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDX25 - DEAD-box helicase 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013264.4 | Intron | NP_037396.3 | ||||
XM_011542790.2 | Intron | XP_011541092.1 | ||||
XM_011542791.2 | Intron | XP_011541093.1 | ||||
XM_017017626.1 | Intron | XP_016873115.1 |
HYLS1 - HYLS1, centriolar and ciliogenesis associated | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PUS3 - pseudouridylate synthase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |