Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCACTTGCTTGGTCTGTGGGTGCC[C/T]GAGTCACTGCCTTGTCTGGAATGAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603287 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PNPO PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PNPO - pyridoxamine 5'-phosphate oxidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRR15L - proline rich 15 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024320.3 | Intron | NP_077296.1 | ||||
XM_005257663.4 | Intron | XP_005257720.1 | ||||
XM_005257664.2 | Intron | XP_005257721.1 | ||||
XM_005257665.1 | Intron | XP_005257722.1 | ||||
XM_005257666.4 | Intron | XP_005257723.1 |