Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGAACTTGCTATACATCAGGAC[C/T]TCCGGTGGGGACCACCGGACTGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300300 MIM: 300356 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BTK - Bruton tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000061.2 | 1894 | Silent Mutation | GAA,GAG | E,E 567 | NP_000052.1 | |
NM_001287344.1 | 1894 | Silent Mutation | GAA,GAG | E,E 601 | NP_001274273.1 | |
NM_001287345.1 | 1894 | Silent Mutation | GAA,GAG | E,E 391 | NP_001274274.1 |
TIMM8A - translocase of inner mitochondrial membrane 8 homolog A (yeast) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |