Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCAGCTCCTCAGCCAGGTCCACG[G/A]GCAGACGGCCCCAGGCATCGCGCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600160 | ||||||||||||||||||||
Literature Links: |
CDKN2A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDKN2A - cyclin dependent kinase inhibitor 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000077.4 | 671 | Missense Mutation | CCC,CTC | P,L 114 | NP_000068.1 | |
NM_001195132.1 | 671 | Missense Mutation | CCC,CTC | P,L 114 | NP_001182061.1 | |
NM_058195.3 | 671 | Silent Mutation | GCC,GCT | A,A 128 | NP_478102.2 | |
NM_058197.4 | 671 | UTR 3 | NP_478104.2 | |||
XM_005251343.1 | 671 | Missense Mutation | CCC,CTC | P,L 63 | XP_005251400.1 | |
XM_011517675.2 | 671 | Missense Mutation | CCC,CTC | P,L 114 | XP_011515977.1 | |
XM_011517676.2 | 671 | Missense Mutation | CCC,CTC | P,L 114 | XP_011515978.1 | |
XM_011517679.1 | 671 | Missense Mutation | CCC,CTC | P,L 63 | XP_011515981.1 |
CDKN2A-AS1 - CDKN2A antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |