Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCGGGCTCACAGTGACCTCCGCC[A/G]CAGGTAAGCGCACGGGGCGGCGCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 131242 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EDN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EDN3 - endothelin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302455.1 | 446 | Missense Mutation | ACA,GCA | T,A 17 | NP_001289384.1 | |
NM_001302456.1 | 446 | Missense Mutation | ACA,GCA | T,A 17 | NP_001289385.1 | |
NM_207032.2 | 446 | Missense Mutation | ACA,GCA | T,A 17 | NP_996915.1 | |
NM_207033.2 | 446 | Missense Mutation | ACA,GCA | T,A 17 | NP_996916.1 | |
NM_207034.2 | 446 | Missense Mutation | ACA,GCA | T,A 17 | NP_996917.1 | |
XM_005260312.4 | 446 | Missense Mutation | ACA,GCA | T,A 17 | XP_005260369.1 | |
XM_005260313.4 | 446 | Missense Mutation | ACA,GCA | T,A 17 | XP_005260370.1 | |
XM_006723734.3 | 446 | Missense Mutation | ACA,GCA | T,A 17 | XP_006723797.1 | |
XM_011528655.2 | 446 | Missense Mutation | ACA,GCA | T,A 17 | XP_011526957.1 |