Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCTGGAGGAGAATCAAGAGCTC[C/T]GAGGTGAGGAAAGAGTCAGGGGATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 305900 MIM: 300248 | ||||||||||||||||||||
Literature Links: |
G6PD PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
G6PD - glucose-6-phosphate dehydrogenase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IKBKG - inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099856.4 | 442 | Nonsense Mutation | CGA,TGA | R,* 130 | NP_001093326.2 | |
NM_001099857.2 | 442 | Nonsense Mutation | CGA,TGA | R,* 62 | NP_001093327.1 | |
NM_001145255.2 | 442 | Nonsense Mutation | CGA,TGA | R,* 62 | NP_001138727.1 | |
NM_001321396.1 | 442 | Nonsense Mutation | CGA,TGA | R,* 62 | NP_001308325.1 | |
NM_001321397.1 | 442 | Nonsense Mutation | CGA,TGA | R,* 62 | NP_001308326.1 | |
NM_003639.4 | 442 | Nonsense Mutation | CGA,TGA | R,* 62 | NP_003630.1 |