Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NOXRED1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NOXRED1 - NADP dependent oxidoreductase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113475.2 | 1678 | UTR 3 | NP_001106946.1 | |||
XM_005267330.3 | 1678 | Intron | XP_005267387.1 | |||
XM_011536427.2 | 1678 | Intron | XP_011534729.1 | |||
XM_011536428.2 | 1678 | Intron | XP_011534730.1 | |||
XM_011536429.2 | 1678 | Intron | XP_011534731.1 | |||
XM_017020969.1 | 1678 | Intron | XP_016876458.1 | |||
XM_017020970.1 | 1678 | Intron | XP_016876459.1 |
SAMD15 - sterile alpha motif domain containing 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |