Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606605 MIM: 615808 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATRIP PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ATRIP - ATR interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271022.1 | Intron | NP_001257951.1 | ||||
NM_001271023.1 | Intron | NP_001257952.1 | ||||
NM_032166.3 | Intron | NP_115542.2 | ||||
NM_130384.2 | Intron | NP_569055.1 |
CCDC51 - coiled-coil domain containing 51 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMA7 - translation machinery associated 7 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |