Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604546 MIM: 611952 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6893 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
MIR6893 - microRNA 6893 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TONSL - tonsoku-like, DNA repair protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013432.4 | 3842 | Missense Mutation | CCC,CTC | P,L 1276 | NP_038460.4 | |
XM_011517048.2 | 3842 | Missense Mutation | CCC,CTC | P,L 952 | XP_011515350.1 | |
XM_011517049.2 | 3842 | Missense Mutation | CCC,CTC | P,L 940 | XP_011515351.1 | |
XM_011517050.2 | 3842 | Intron | XP_011515352.1 |
TONSL-AS1 - TONSL antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VPS28 - VPS28, ESCRT-I subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |