Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 603454 | |||||||||||||||||||||||
Literature Links: |
CRADD PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
CRADD - CASP2 and RIPK1 domain containing adaptor with death domain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320099.1 | 186 | Missense Mutation | AAG,GAG | K,E 19 | NP_001307028.1 | |
NM_001320100.1 | 186 | Missense Mutation | AAG,GAG | K,E 19 | NP_001307029.1 | |
NM_001320101.1 | 186 | Missense Mutation | AAG,GAG | K,E 19 | NP_001307030.1 | |
NM_003805.4 | 186 | Missense Mutation | AAG,GAG | K,E 19 | NP_003796.1 | |
XM_017020142.1 | 186 | Missense Mutation | AAG,GAG | K,E 19 | XP_016875631.1 | |
XM_017020143.1 | 186 | Missense Mutation | AAG,GAG | K,E 19 | XP_016875632.1 | |
XM_017020144.1 | 186 | Missense Mutation | AAG,GAG | K,E 19 | XP_016875633.1 | |
XM_017020145.1 | 186 | Missense Mutation | AAG,GAG | K,E 19 | XP_016875634.1 |