Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606883 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IRAK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PUS7L - pseudouridylate synthase 7 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098614.2 | 192 | Intron | NP_001092084.1 | |||
NM_001098615.1 | 192 | Intron | NP_001092085.1 | |||
NM_001271826.1 | 192 | Intron | NP_001258755.1 | |||
NM_031292.4 | 192 | Intron | NP_112582.3 | |||
XM_006719623.2 | 192 | Intron | XP_006719686.1 | |||
XM_011538790.2 | 192 | Intron | XP_011537092.1 | |||
XM_011538791.2 | 192 | Intron | XP_011537093.1 | |||
XM_017019996.1 | 192 | Intron | XP_016875485.1 | |||
XM_017019997.1 | 192 | Intron | XP_016875486.1 | |||
XM_017019998.1 | 192 | Intron | XP_016875487.1 |