Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608150 MIM: 610750 | ||||||||||||||||||||
Literature Links: |
PPHLN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPHLN1 - periphilin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143787.1 | Intron | NP_001137259.1 | ||||
NM_001143788.1 | Intron | NP_001137260.1 | ||||
NM_001143789.1 | Intron | NP_001137261.1 | ||||
NM_016488.6 | Intron | NP_057572.5 | ||||
NM_201438.1 | Intron | NP_958846.1 | ||||
NM_201439.1 | Intron | NP_958847.1 | ||||
NM_201440.1 | Intron | NP_958848.1 | ||||
NM_201515.1 | Intron | NP_958923.1 |
ZCRB1 - zinc finger CCHC-type and RNA binding motif containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |