Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300506 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TSC22D3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TSC22D3 - TSC22 domain family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001015881.1 | Intron | NP_001015881.1 | ||||
NM_001318468.1 | Intron | NP_001305397.1 | ||||
NM_001318470.1 | Intron | NP_001305399.1 | ||||
NM_004089.3 | Intron | NP_004080.2 | ||||
NM_198057.2 | Intron | NP_932174.1 | ||||
XM_005262099.1 | Intron | XP_005262156.1 | ||||
XM_005262100.1 | Intron | XP_005262157.1 | ||||
XM_005262102.1 | Intron | XP_005262159.1 | ||||
XM_005262103.3 | Intron | XP_005262160.1 | ||||
XM_011530884.1 | Intron | XP_011529186.1 | ||||
XM_017029335.1 | Intron | XP_016884824.1 |