Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604638 MIM: 608839 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTN4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ACTN4 - actinin alpha 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322033.1 | Intron | NP_001308962.1 | ||||
NM_004924.5 | Intron | NP_004915.2 | ||||
XM_005259281.4 | Intron | XP_005259338.1 | ||||
XM_006723406.2 | Intron | XP_006723469.1 | ||||
XM_017027331.1 | Intron | XP_016882820.1 |
CAPN12 - calpain 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144691.4 | Intron | NP_653292.2 | ||||
XM_017026355.1 | Intron | XP_016881844.1 | ||||
XM_017026356.1 | Intron | XP_016881845.1 | ||||
XM_017026357.1 | Intron | XP_016881846.1 | ||||
XM_017026358.1 | Intron | XP_016881847.1 | ||||
XM_017026359.1 | Intron | XP_016881848.1 | ||||
XM_017026360.1 | Intron | XP_016881849.1 | ||||
XM_017026361.1 | Intron | XP_016881850.1 | ||||
XM_017026362.1 | Intron | XP_016881851.1 | ||||
XM_017026363.1 | Intron | XP_016881852.1 | ||||
XM_017026364.1 | Intron | XP_016881853.1 | ||||
XM_017026365.1 | Intron | XP_016881854.1 |
Set Membership: |
HapMap |