Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCTCTCATACAGGTGCAGTTTGT[A/C]TTTGTTGGAGTGGAGCATCTTGACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607362 | ||||||||||||||||||||
Literature Links: |
RGMA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RGMA - repulsive guidance molecule family member a | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166283.1 | 1480 | Missense Mutation | GAC,TAC | D,Y 417 | NP_001159755.1 | |
NM_001166286.1 | 1480 | Missense Mutation | GAC,TAC | D,Y 393 | NP_001159758.1 | |
NM_001166287.1 | 1480 | Missense Mutation | GAC,TAC | D,Y 393 | NP_001159759.1 | |
NM_001166288.1 | 1480 | Missense Mutation | GAC,TAC | D,Y 393 | NP_001159760.1 | |
NM_001166289.1 | 1480 | Missense Mutation | GAC,TAC | D,Y 393 | NP_001159761.1 | |
NM_020211.2 | 1480 | Missense Mutation | GAC,TAC | D,Y 409 | NP_064596.2 |