Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613066 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KRT18P59 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KRT18P59 - keratin 18 pseudogene 59 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PKNOX2 - PBX/knotted 1 homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022062.2 | 921 | Intron | NP_071345.2 | |||
XM_005271642.2 | 921 | Intron | XP_005271699.1 | |||
XM_005271643.2 | 921 | Intron | XP_005271700.1 | |||
XM_006718894.2 | 921 | Intron | XP_006718957.1 | |||
XM_011542944.2 | 921 | Intron | XP_011541246.1 | |||
XM_011542945.2 | 921 | Intron | XP_011541247.1 | |||
XM_011542946.1 | 921 | Intron | XP_011541248.1 | |||
XM_011542947.2 | 921 | Intron | XP_011541249.1 | |||
XM_017018110.1 | 921 | UTR 5 | XP_016873599.1 | |||
XM_017018111.1 | 921 | Intron | XP_016873600.1 | |||
XM_017018112.1 | 921 | Intron | XP_016873601.1 |
TMEM218 - transmembrane protein 218 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |