Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCTTTGTTCCTCTTATGACGTTG[C/T]TCTTTCTGAACTTCCTAAAGCCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610841 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105374546 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC105374546 - uncharacterized LOC105374546 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STIM2 - stromal interaction molecule 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001169117.1 | Intron | NP_001162588.1 | ||||
NM_001169118.1 | Intron | NP_001162589.1 | ||||
NM_020860.3 | Intron | NP_065911.3 |
TBC1D19 - TBC1 domain family member 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |