Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||
Literature Links: |
FAM133A PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
FAM133A - family with sequence similarity 133 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171109.1 | 210 | Intron | NP_001164580.1 | |||
NM_001171110.1 | 210 | Intron | NP_001164581.1 | |||
NM_001171111.1 | 210 | Intron | NP_001164582.1 | |||
NM_173698.2 | 210 | Intron | NP_775969.1 | |||
XM_006724641.3 | 210 | UTR 5 | XP_006724704.1 | |||
XM_011530925.2 | 210 | Intron | XP_011529227.1 | |||
XM_011530926.2 | 210 | Intron | XP_011529228.1 | |||
XM_011530927.2 | 210 | Intron | XP_011529229.1 | |||
XM_011530928.1 | 210 | Intron | XP_011529230.1 | |||
XM_011530929.1 | 210 | Intron | XP_011529231.1 | |||
XM_011530931.2 | 210 | UTR 5 | XP_011529233.1 | |||
XM_017029454.1 | 210 | Intron | XP_016884943.1 | |||
XM_017029455.1 | 210 | Intron | XP_016884944.1 | |||
XM_017029456.1 | 210 | Intron | XP_016884945.1 |
Set Membership: |
HapMap |