Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601591 | ||||||||||||||||||||
Literature Links: |
PRKG2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PRKG2 - protein kinase, cGMP-dependent, type II | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282480.1 | Intron | NP_001269409.1 | ||||
NM_001282481.1 | Intron | NP_001269410.1 | ||||
NM_001282482.1 | Intron | NP_001269411.1 | ||||
NM_001282483.1 | Intron | NP_001269412.1 | ||||
NM_001282485.1 | Intron | NP_001269414.1 | ||||
NM_006259.2 | Intron | NP_006250.1 |