Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614183 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DIS3L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DIS3L - DIS3 like exosome 3'-5' exoribonuclease | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143688.2 | Intron | NP_001137160.1 | ||||
NM_001323936.1 | Intron | NP_001310865.1 | ||||
NM_001323937.1 | Intron | NP_001310866.1 | ||||
NM_001323938.1 | Intron | NP_001310867.1 | ||||
NM_001323939.1 | Intron | NP_001310868.1 | ||||
NM_001323940.1 | Intron | NP_001310869.1 | ||||
NM_001323941.1 | Intron | NP_001310870.1 | ||||
NM_001323943.1 | Intron | NP_001310872.1 | ||||
NM_001323944.1 | Intron | NP_001310873.1 | ||||
NM_001323945.1 | Intron | NP_001310874.1 | ||||
NM_001323946.1 | Intron | NP_001310875.1 | ||||
NM_001323948.1 | Intron | NP_001310877.1 | ||||
NM_133375.4 | Intron | NP_588616.1 | ||||
XM_005254146.3 | Intron | XP_005254203.1 |
Set Membership: |
HapMap |