Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600844 MIM: 174762 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
P2RX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
P2RX2 - purinergic receptor P2X 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLE - DNA polymerase epsilon, catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006231.3 | 6863 | Intron | NP_006222.2 | |||
XM_011534795.2 | 6863 | Missense Mutation | CAG,CGG | Q,R 2220 | XP_011533097.1 | |
XM_011534797.2 | 6863 | Missense Mutation | CAG,CGG | Q,R 1913 | XP_011533099.1 | |
XM_011534798.1 | 6863 | Missense Mutation | CAG,CGG | Q,R 1774 | XP_011533100.1 | |
XM_011534799.2 | 6863 | Intron | XP_011533101.1 | |||
XM_011534800.2 | 6863 | Intron | XP_011533102.1 | |||
XM_011534802.2 | 6863 | Missense Mutation | CAG,CGG | Q,R 1216 | XP_011533104.1 |
Set Membership: |
HapMap |