Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606992 MIM: 606671 | ||||||||||||||||||||
Literature Links: |
IP6K2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IP6K2 - inositol hexakisphosphate kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005909.2 | Intron | NP_001005909.1 | ||||
NM_001005910.2 | Intron | NP_001005910.1 | ||||
NM_001005911.2 | Intron | NP_001005911.1 | ||||
NM_001146178.2 | Intron | NP_001139650.1 | ||||
NM_001146179.2 | Intron | NP_001139651.1 | ||||
NM_001190316.1 | Intron | NP_001177245.1 | ||||
NM_001190317.1 | Intron | NP_001177246.1 | ||||
NM_016291.3 | Intron | NP_057375.2 |
NCKIPSD - NCK interacting protein with SH3 domain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |