Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610613 | ||||||||||||||||||||||||||||||||
Literature Links: |
CYP11B1 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian
|
CEPH (CEU) - Not Available | |||
EAS - Not Available | African American
|
YRI (Yoruba) - Not Available | |||
SAS - Not Available | Japanese
|
CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Chinese
|
JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CYP11B1 - cytochrome P450 family 11 subfamily B member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000497.3 | 2044 | UTR 3 | NP_000488.3 | |||
NM_001026213.1 | 2044 | UTR 3 | NP_001021384.1 | |||
XM_011516874.2 | 2044 | UTR 3 | XP_011515176.1 | |||
XM_011516875.1 | 2044 | UTR 3 | XP_011515177.1 | |||
XM_017013146.1 | 2044 | UTR 3 | XP_016868635.1 |