Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 191315 MIM: 604514 | ||||||||||||||||||||
Literature Links: |
NTRK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NTRK1 - neurotrophic receptor tyrosine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007792.1 | 72 | Intron | NP_001007793.1 | |||
NM_001012331.1 | 72 | Intron | NP_001012331.1 | |||
NM_002529.3 | 72 | Intron | NP_002520.2 |
SH2D2A - SH2 domain containing 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161441.1 | 72 | UTR 5 | NP_001154913.1 | |||
NM_001161442.1 | 72 | UTR 5 | NP_001154914.1 | |||
NM_001161443.1 | 72 | Intron | NP_001154915.1 | |||
NM_001161444.1 | 72 | UTR 5 | NP_001154916.1 | |||
NM_003975.3 | 72 | UTR 5 | NP_003966.2 |