Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTLL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TTLL4 - tubulin tyrosine ligase like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014640.4 | Intron | NP_055455.3 | ||||
XM_006712875.2 | Intron | XP_006712938.1 | ||||
XM_017005387.1 | Intron | XP_016860876.1 | ||||
XM_017005388.1 | Intron | XP_016860877.1 | ||||
XM_017005389.1 | Intron | XP_016860878.1 | ||||
XM_017005390.1 | Intron | XP_016860879.1 | ||||
XM_017005391.1 | Intron | XP_016860880.1 | ||||
XM_017005392.1 | Intron | XP_016860881.1 |