Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607337 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BEST3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BEST3 - bestrophin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282613.1 | Intron | NP_001269542.1 | ||||
NM_001282614.1 | Intron | NP_001269543.1 | ||||
NM_001282615.1 | Intron | NP_001269544.1 | ||||
NM_001282616.1 | Intron | NP_001269545.1 | ||||
NM_032735.2 | Intron | NP_116124.2 | ||||
NM_152439.3 | Intron | NP_689652.2 | ||||
XM_006719256.3 | Intron | XP_006719319.1 | ||||
XM_011537961.1 | Intron | XP_011536263.1 | ||||
XM_011537962.2 | Intron | XP_011536264.1 | ||||
XM_011537963.1 | Intron | XP_011536265.1 | ||||
XM_011537965.2 | Intron | XP_011536267.1 | ||||
XM_017018865.1 | Intron | XP_016874354.1 |