Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607700 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF40 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RNF40 - ring finger protein 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207033.1 | 2327 | Intron | NP_001193962.1 | |||
NM_001207034.1 | 2327 | Intron | NP_001193963.1 | |||
NM_001286572.2 | 2327 | Intron | NP_001273501.1 | |||
NM_014771.3 | 2327 | Intron | NP_055586.1 | |||
XM_011545997.1 | 2327 | Intron | XP_011544299.1 |
ZNF629 - zinc finger protein 629 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080417.1 | 2327 | Missense Mutation | CCC,GCC | P,A 707 | NP_001073886.1 | |
XM_005255222.3 | 2327 | Missense Mutation | CCC,GCC | P,A 693 | XP_005255279.1 |
Set Membership: |
HapMap |