Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
56 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610816 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC25A33 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC25A33 - solute carrier family 25 member 33 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM201 - transmembrane protein 201 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010866.3 | Intron | NP_001010866.1 | ||||
NM_001130924.2 | Intron | NP_001124396.2 | ||||
XM_006710417.3 | Intron | XP_006710480.1 | ||||
XM_011540910.2 | Intron | XP_011539212.1 | ||||
XM_017000549.1 | Intron | XP_016856038.1 | ||||
XM_017000550.1 | Intron | XP_016856039.1 | ||||
XM_017000551.1 | Intron | XP_016856040.1 | ||||
XM_017000552.1 | Intron | XP_016856041.1 |
Set Membership: |
HapMap |