Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAGGGAGGAGGTGAAGCTGAGGC[A/G]AGGCTGCTTCTTACTCAGCTGCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602038 MIM: 148022 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUSP8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DUSP8 - dual specificity phosphatase 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KRTAP5-1 - keratin associated protein 5-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KRTAP5-AS1 - KRTAP5-1/KRTAP5-2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |