Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612815 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZDHHC13 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZDHHC13 - zinc finger DHHC-type containing 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001483.2 | Intron | NP_001001483.1 | ||||
NM_019028.2 | Intron | NP_061901.2 | ||||
XM_005252996.2 | Intron | XP_005253053.1 | ||||
XM_011520193.1 | Intron | XP_011518495.1 | ||||
XM_011520194.2 | Intron | XP_011518496.1 | ||||
XM_011520195.1 | Intron | XP_011518497.1 |
Set Membership: |
HapMap |