Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTGGAGAGAAAGGGGGAAAAAAA[A/C]CCACTCTTGCATTAAATCCTGGTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C10orf71 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C10orf71 - chromosome 10 open reading frame 71 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135196.1 | Intron | NP_001128668.1 | ||||
XM_005269476.4 | Intron | XP_005269533.1 | ||||
XM_005269477.3 | Intron | XP_005269534.1 | ||||
XM_005269478.4 | Intron | XP_005269535.1 | ||||
XM_017015619.1 | Intron | XP_016871108.1 |
C10orf71-AS1 - C10orf71 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |