Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 606990 MIM: 605810 | |||||||||||||||||||||||
Literature Links: |
COPB2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
COPB2 - coatomer protein complex subunit beta 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPS22 - mitochondrial ribosomal protein S22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020191.2 | Intron | NP_064576.1 | ||||
XM_005247640.2 | Intron | XP_005247697.1 | ||||
XM_006713703.3 | Intron | XP_006713766.1 | ||||
XM_011512995.2 | Intron | XP_011511297.1 | ||||
XM_011512996.2 | Intron | XP_011511298.1 |