Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 146660 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IL7 - interleukin 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000880.3 | Intron | NP_000871.1 | ||||
NM_001199886.1 | Intron | NP_001186815.1 | ||||
NM_001199887.1 | Intron | NP_001186816.1 | ||||
NM_001199888.1 | Intron | NP_001186817.1 | ||||
XM_011517522.2 | Intron | XP_011515824.1 | ||||
XM_011517523.2 | Intron | XP_011515825.1 | ||||
XM_017013397.1 | Intron | XP_016868886.1 |
ZC2HC1A - zinc finger C2HC-type containing 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016010.2 | Intron | NP_057094.2 | ||||
XM_005251256.1 | Intron | XP_005251313.1 | ||||
XM_006716454.2 | Intron | XP_006716517.1 | ||||
XM_011517539.2 | Intron | XP_011515841.1 |
Set Membership: |
HapMap |