Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615392 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SFMBT2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SFMBT2 - Scm-like with four mbt domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018039.1 | 3108 | UTR 3 | NP_001018049.1 | |||
NM_001029880.2 | 3108 | UTR 3 | NP_001025051.1 | |||
XM_005252551.2 | 3108 | UTR 3 | XP_005252608.1 | |||
XM_006717490.2 | 3108 | UTR 3 | XP_006717553.1 | |||
XM_011519611.1 | 3108 | UTR 3 | XP_011517913.1 | |||
XM_017016466.1 | 3108 | UTR 3 | XP_016871955.1 |
Set Membership: |
HapMap |