Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP112 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CEP112 - centrosomal protein 112 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037325.2 | Intron | NP_001032402.1 | ||||
NM_001199165.2 | Intron | NP_001186094.1 | ||||
NM_001302891.1 | Intron | NP_001289820.1 | ||||
XM_005257119.4 | Intron | XP_005257176.1 | ||||
XM_005257125.3 | Intron | XP_005257182.1 | ||||
XM_005257126.4 | Intron | XP_005257183.1 | ||||
XM_006721740.2 | Intron | XP_006721803.1 | ||||
XM_006721744.3 | Intron | XP_006721807.1 | ||||
XM_011524461.2 | Intron | XP_011522763.1 | ||||
XM_011524462.2 | Intron | XP_011522764.1 | ||||
XM_011524463.2 | Intron | XP_011522765.1 | ||||
XM_011524464.2 | Intron | XP_011522766.1 | ||||
XM_011524465.2 | Intron | XP_011522767.1 | ||||
XM_011524466.2 | Intron | XP_011522768.1 | ||||
XM_011524467.2 | Intron | XP_011522769.1 | ||||
XM_017024302.1 | Intron | XP_016879791.1 | ||||
XM_017024303.1 | Intron | XP_016879792.1 |