Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF668 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF668 - zinc finger protein 668 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172668.1 | 1953 | Nonsense Mutation | CAG,TAG | Q,* 548 | NP_001166139.1 | |
NM_001172669.1 | 1953 | Nonsense Mutation | CAG,TAG | Q,* 571 | NP_001166140.1 | |
NM_001172670.1 | 1953 | Nonsense Mutation | CAG,TAG | Q,* 548 | NP_001166141.1 | |
NM_024706.4 | 1953 | Nonsense Mutation | CAG,TAG | Q,* 548 | NP_078982.3 |