Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610305 MIM: 601607 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DERL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DERL3 - derlin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002862.2 | Intron | NP_001002862.1 | ||||
NM_001135751.1 | Intron | NP_001129223.1 | ||||
NM_198440.3 | Intron | NP_940842.2 | ||||
XM_011530505.2 | Intron | XP_011528807.1 | ||||
XM_011530506.2 | Intron | XP_011528808.1 | ||||
XM_017029078.1 | Intron | XP_016884567.1 | ||||
XM_017029079.1 | Intron | XP_016884568.1 | ||||
XM_017029080.1 | Intron | XP_016884569.1 | ||||
XM_017029081.1 | Intron | XP_016884570.1 | ||||
XM_017029082.1 | Intron | XP_016884571.1 |
SMARCB1 - SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |