Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 173325 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
JUP PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
JUP - junction plakoglobin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002230.2 | 2993 | UTR 3 | NP_002221.1 | |||
NM_021991.2 | 2993 | UTR 3 | NP_068831.1 | |||
XM_006721873.2 | 2993 | UTR 3 | XP_006721936.1 | |||
XM_006721874.2 | 2993 | UTR 3 | XP_006721937.1 | |||
XM_006721875.1 | 2993 | UTR 3 | XP_006721938.1 | |||
XM_006721878.1 | 2993 | UTR 3 | XP_006721941.1 | |||
XM_011524753.2 | 2993 | UTR 3 | XP_011523055.1 | |||
XM_011524755.1 | 2993 | UTR 3 | XP_011523057.1 | |||
XM_011524756.1 | 2993 | UTR 3 | XP_011523058.1 | |||
XM_011524757.2 | 2993 | UTR 3 | XP_011523059.1 | |||
XM_011524758.1 | 2993 | UTR 3 | XP_011523060.1 | |||
XM_017024588.1 | 2993 | UTR 3 | XP_016880077.1 | |||
XM_017024589.1 | 2993 | UTR 3 | XP_016880078.1 | |||
XM_017024590.1 | 2993 | UTR 3 | XP_016880079.1 |